The Sklar Black™ joseph skin hook is a handheld retractor commonly used for holding back the edges of skin during laser intranasal and pharyngeal procedures.
- Safe and durable coating
- Reduces laser reflection to less than one-tenth of a percent
It is available with a single or dual prong, sharp hook. The handle is often round and knurled and it tapers to the hook end. Sklar Black™ Laser instruments feature a non-reflective black polymer coating applied over the sandblasted instrument developed specifically for laser surgeries.
It ensures a safer surgical environment for patients and surgical staff. Sklar Black™ laser coatings are guaranteed for one year against manufacturing defects.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.