Water Accelerometer | Avantor
Water Accelerometer
Water Accelerometer
Certificates
Water Accelerometer
Water Accelerometer
2 Options Available Below
Product Details & Documents
Visualize acceleration and centripetal force with this unique device.
  • Dramatic visual demonstration of accelerated vs equilibrium motion
  • Liquid surface shows relative acceleration of a moving system
  • Printed grid allows for quntitative evaluation of slope
  • Turntable provides easy method for studying rotating systems
  • Student lab sheet provided
The unit is comprised of a clear rectangular container, marked with a centimeter grid, and partially filled with water. At equilibrium, the water forms a level, horizontal line on the grid. When the device is accelerated, the slope of the water level changes, allowing a quantitative analysis of Newton’s 2nd Law. Affix to a dynamics cart to compare acceleration down an inclined plane at different angles. Or, visualize the acceleration of a half-atwood machine under different conditions. The unit can also be attached to a turntable, allowing students to study the centrifugal effects of rotation.
Ordering information: Accelerometer and turntable available separately. Unit shipped dry; water not included. Recommended: Use colored water to enhance the visual impact for a demonstration. Use a camera to accurately capture and evaluate the slope.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Water accelerometer only
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $40.95
    $40.95
    Liquidation
    Log in pour la disponibilité
    Case of 12
    Case of 12 - $491.55
    $491.55
    Liquidation
    Each
       $40.95
    Case of 12
       $491.55
    Supplier Number: PH10407
  • Turn table for accelerometer
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $81.85
    $81.85
    Liquidation
    Log in pour la disponibilité
    Case of 6
    Case of 6 - $491.10
    $491.10
    Liquidation
    Each
       $81.85
    Case of 6
       $491.10
    Supplier Number: PH10407T