Mix & Go!™ E. coli Transformation Kit and Buffer Set
À propos de cet article
The Mix & Go!™ E. coli transformation kit and Mix & Go!™ E. coli transformation buffer set are convenient methods for the preparation of competent E. coli cells for simple and highly efficient DNA transformation.
It is very simple and efficient DNA transformation
- It completely eliminates the requirement for heat shocking and related procedures
- Transformation efficiencies are typically on the order of 108 to 109 transformants/µg plasmid DNA
The Mix & Go! method completely eliminates the requirement for heat shocking and related procedures. Instead, Mix & Go! bacterial transformation can be performed by adding DNA to Mix & Go! competent cells and spreading onto a plate. Transformation efficiencies are typically on the order of 108 to 109 transformants/µg plasmid DNA with most E. coli strains. These formulated reagents make it easy to generate Mix & Go! competent cells from current E. coli strains that are available in the laboratory. Simply grow the E. coli strain of your choice, wash, then resuspend the cells in the provided buffers. The cells are now transformation ready. The Mix & Go! E. coli Transformation Kit includes all buffers and ZymoBroth medium to generate 20 ml of Mix & Go! competent cells. The Mix & Go! E. coli Transformation Buffer Set includes all buffers that are required to generate 60 ml of Mix & Go! competent cells, and the medium (broth) is supplied by the user.
2 Options Available Below
- L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
- Return Policy
Product Details & Documents
The Mix & Go!™ E. coli transformation kit and Mix & Go!™ E. coli transformation buffer set are convenient methods for the preparation of competent E. coli cells for simple and highly efficient DNA transformation.
It is very simple and efficient DNA transformation
- It completely eliminates the requirement for heat shocking and related procedures
- Transformation efficiencies are typically on the order of 108 to 109 transformants/µg plasmid DNA
The Mix & Go! method completely eliminates the requirement for heat shocking and related procedures. Instead, Mix & Go! bacterial transformation can be performed by adding DNA to Mix & Go! competent cells and spreading onto a plate. Transformation efficiencies are typically on the order of 108 to 109 transformants/µg plasmid DNA with most E. coli strains. These formulated reagents make it easy to generate Mix & Go! competent cells from current E. coli strains that are available in the laboratory. Simply grow the E. coli strain of your choice, wash, then resuspend the cells in the provided buffers. The cells are now transformation ready. The Mix & Go! E. coli Transformation Kit includes all buffers and ZymoBroth medium to generate 20 ml of Mix & Go! competent cells. The Mix & Go! E. coli Transformation Buffer Set includes all buffers that are required to generate 60 ml of Mix & Go! competent cells, and the medium (broth) is supplied by the user.
Product Family Options
Informations sur le produit
- DésignationTailleAvailabilityPrice
- Mix & Go!™ E. coli transformation kitup to 20 mlEach$220.68Kit of 1$220.68Supplier Number: T3001L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
- Mix & Go!™ E. coli Transformation Buffer Kitup to 60 mlEach$319.12KIT(1items)$243.54Kit of 1$319.12Supplier Number: T3002L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.