ContactUsLinkComponent
Air Core Solenoid
Catalogue # 470219-586
Fournisseur:  Shiv Dial Sud & Sons
undefined
Air Core Solenoid
Catalogue # 470219-586
Fournisseur:  Shiv Dial Sud & Sons
Supplier Number:  PH50255
Spécifications
  • Description:
    Air Core Solenoid
  • Height:
    7.25"
  • Cat. no.:
    470219-586
  • Supplier No.:
    PH50255
  • Diameter:
    2.5"
Spécifications
À propos de cet article
The Air Core Solenoid is perfect for induced magnetic field studies.
  • Solenoid Can Also Work with a Current Balance
  • Coils Wound on a Plastic Tube
  • Copper Wire Has 575 Turns
  • Solenoid Carries 5A Current Continuously
  • For Induced Magnetic Field Studies
It can also work with a current balance to measure the force on a conductor in a magnetic field. The solenoid has coils wound on a plastic tube with approximately 575 turns of copper wire.
It can carry a current of 5A continuously or 10A intermittently.
Resistance is - 1.2 ohm
Inductance is – 6 mH