Friction Board
Friction Board
Catalog # 470219-584
Supplier: Shiv Dial Sud & Sons
Fournisseur #: PH10568
Jump to:
Product Details & Documents
To Study the effects of friction using 4 different surface materials on a board.
- Introduce friction
- Includes four surface materials
- Instructions included
Includes surface board, hooks, and wooden blocks, with instruction manual.
Spécifications
- Désignation:Friction Board