À propos de cet article
The Sklar Blue™ Steiner-Auvard Weighted Vaginal Speculum is a single end vaginal speculum commonly used in OB/GYN procedures such as vaginal hysterectomy or dilation and curettage.
- It is weighted with a medium, 21/2 lbs.
Sklar Blue™ electrosurgical instruments feature high quality stainless steel with an insulated coating. This safe coating prevents radio frequency currents from being transmitted through the instrument which reduces the burning of non-targeted tissue. Sklar Blue™ instruments are fully autoclavable and may also be cleaned ultrasonically. Sklar Blue™ electrosurgical coatings are guaranteed for one year against manufacturing defects.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.