SLIKSTOPPER™ Mat, Brady | Avantor
SLIKSTOPPER™ Mat, Brady
Drain Cover
Catalog # 10809-772
Supplier:   Brady Worldwide
Fournisseur #:  107768
SLIKSTOPPER™ Mat, Brady
Catalog # 10809-772
Supplier:  Brady Worldwide
SLIKSTOPPER™ Mat, Brady
SLIKSTOPPER™ Mat, Brady
Drain Cover
Price
$967.35
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Roll of 1
Product Details & Documents
The SLIKSTOPPER™ is made from chemical-resistant, flexible PVC, this mat seals drains tightly to prevent spills from spreading
Resists water, oil and most aggressive fluids. Easy application and soap and water cleanup for re-use. Vinyl material. Yellow color.
Spécifications
  • Désignation:
    Drain Cover
  • Cat. No.:
    10809-772
  • Dimensions:
    24W x 24L"