Microscope Slide Coverslips, Polysciences | Avantor
Microscope Slide Coverslips, Polysciences
Supplier:   Polysciences Inc
Microscope Slide Coverslips, Polysciences
Supplier:  Polysciences Inc
Certificates
Microscope Slide Coverslips, Polysciences
Microscope Slide Coverslips, Polysciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Rectangular glass coverslips made of Borosilicate glass. Ideal coverslip for various methods, including routine and special stains, immunohistochemistry, and in situ hybridization.
Product Family Options
Informations sur le produit
  • Désignation
    L×l
    Épaisseur
    Availability
    Price
  • Glass cover slips, rectangular
    30×22 mm
    17 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $187.68
    $187.68
    Pack of 1
       $187.68
    Supplier Number: 23999-1
  • Glass cover slips, rectangular
    40×24 mm
    17 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $194.59
    $194.59
    Pack of 1
       $194.59
    Supplier Number: 24070-1