Needle Electroscope, Large
Needle Electroscope, Large
Catalog # 470219-526
Supplier:   Wards
 :  PH0916
Needle Electroscope, Large
Catalog # 470219-526
Supplier:  Wards
Needle Electroscope, Large
Needle Electroscope, Large
Needle Electroscope, Large
Price
$146.25
Log in for availability
Each
Product Details & Documents
This extra large electroscope is ideal for classroom demonstrations of electrostatic principles. A lightweight metal vane is used as a pointer in the electroscope. This lightweight metal blade deflects when the electroscope plate is charged. A transparent scale is provided to measure the angle of the deflection. Front glass window slides up to allow insertion of ionizing material in chamber.
This electroscope comes with two accessories: an electrophorus disc and a spherical electrode.
Assembled, the electroscope measures 9.75 x 8 x 4.75".
Specifications
  • Désignation:
    Needle Electroscope, Large