Mounted Mirror | Avantor
Mounted Mirror
Supplier:   Wards
Mounted Mirror
Supplier:  Wards
Certificates
Mounted Mirror
2 Options Available Below
Product Details & Documents
No need for a mirror support stand with these mirrors. Silvered back mirror with a protective coating. Mounted on wooden block 20mm thick.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Mounted mirror, 75x25 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4.60
    $4.60
    Liquidation
    Each
       $4.60
    Supplier Number: PH0515A
  • Mounted mirror, 150x50 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $16.75
    $16.75
    Each
       $16.75
    Supplier Number: PH0515E