ContactUsLinkComponent
Truss Kit
  470219-430
 :  Wards
undefined
Truss Kit
  470219-430
 :  Wards
 :  PH0344SFT
  • Description:
    Truss Kit
  • Cat. no.:
    470219-430
  • Supplier No.:
    PH0344SFT
Studying dynamic tension has never been easier with this simple form truss kit. The kit includes a half-meter truss with hooks on both sides, V-shaped clamp.Recommended stand and rod, item 470137-892 Required but not included are masses and spring scale.