ContactUsLinkComponent
Displacement Vessel
  470219-414
 :  Wards
undefined
Displacement Vessel
  470219-414
 :  Wards
 :  PH0119C
  • Description:
    Displacement Vessel
  • Cat. no.:
    470219-414
  • Supplier No.:
    PH0119C
A perfect tool for studying density, specific gravity, and water displacement. The 250mL polypropylene overflow can with spout only requires a catch bucket for the overflow.