Vernier® pH Buffer Capsules, 3×10 | Avantor
Vernier® pH Buffer Capsules, 3×10
Vernier pH buffer capsules, 3×10
Catalog # CA470219-362
New Product
Fournisseur #:  PH-BUFCAP
Vernier® pH Buffer Capsules, 3×10
Catalog # CA470219-362
New Product
CAS Number:  
Vernier® pH Buffer Capsules, 3×10
Vernier® pH Buffer Capsules, 3×10
Vernier pH buffer capsules, 3×10
CAS Number:  
Price
$70.09
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The pH Buffer Capsules (3x10) kit is a convenient set of three vials containing capsules to make pH 4, pH 7, and pH 10 buffer standards for calibrating pH sensors. Each vial contains 10 capsules, each of which produces 100 mL of solution when mixed with DI water. Add three drops of the buffer preservative to 100 mL of your buffer solution to prolong the shelf life. Hydrion Color Key Buffer Preservative is a combination mold inhibitor and universal indicator for color coding the buffer solutions: orange for pH 4, green for pH 7, and blue for pH 10.
Spécifications
Spécifications
  • Désignation:
    Vernier pH buffer capsules, 3×10