Flame Resistant Long Sleeve Henley Shirt with Max Comfort™ Material, Navy Blue, MCR Safety
Supplier: MCR Safety
Certificates
About this item
Whether working in the rugged oil and gas industry or spending the day working near electrical hazards, stay safe and comfortable in MCR Safety's Flame Resistant (FR) Gear made with Max Comfort™ fabrics
- 100% FR Cotton
- Left chest pocket
- Moisture wicking and breathable
- Rib-knit collar and cuff
- NFPA 2112, NFPA 70E, UL Certified
The H1 Henley Shirt features a 100% FR cotton material for maximum comfort. The Max Comfort™ material meets the NFPA 2112-2012 Standard for Flame Resistant Garments and NFPA 70E Standard for Electrical Safety in the Workplace. All of MCR Safety's FR Garments are UL Certified and have a lifetime FR guarantee.
Certifications: NFPA 2112, NFPA 70E, UL Certified.
Specifications
- Catalog No:
- 10813-476
- 10813-474
- 10813-472
- 10813-470
- 10813-484
- 10813-482
- 10813-480
- 10813-478
- Garment application:
- Industrial, Fire protection, Oils, fats and fuels, General Purpose
- Industrial, Fire protection, Oils, fats and fuels, General Purpose
- Industrial, Fire protection, Oils, fats and fuels, General Purpose
- Industrial, Fire protection, Oils, fats and fuels, General Purpose
- Industrial, Fire protection, Oils, fats and fuels, General Purpose
- Industrial, Fire protection, Oils, fats and fuels, General Purpose
- Industrial, Fire protection, Oils, fats and fuels, General Purpose
- Industrial, Fire protection, Oils, fats and fuels, General Purpose
- Risk area:
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Gender:
- For Men
- For Men
- For Men
- For Men
- For Men
- For Men
- For Men
- For Men
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Fabric:
- 100% FR cotton
- 100% FR cotton
- 100% FR cotton
- 100% FR cotton
- 100% FR cotton
- 100% FR cotton
- 100% FR cotton
- 100% FR cotton
- Thickness:
- 7 oz.
- 7 oz.
- 7 oz.
- 7 oz.
- 7 oz.
- 7 oz.
- 7 oz.
- 7 oz.