Caps, BASIK™, MicroSolv
Caps, BASIK™, MicroSolv
Caps, BASIK™, MicroSolv
Caps, BASIK™, MicroSolv
Caps, BASIK™, MicroSolv
For use with 12 x 32 mm autosampler vials.
Product Family Options
Snap/Crimp Top
  • Matière
    Septum
    Taille du bouchon
    Cap Color
    Cap Type
  • PE
    Soft-Guard, White Silicone Rubber / Red PTFE
    11 mm
    Blue
    Snap cap
    Case of 1,000
    Case of 1,000 - $530.20
    $530.20
    Case of 1,000
       $530.20
     : 9502C-E-10B-M
  • PE
    Soft-Guard, White Silicone Rubber / Red PTFE
    11 mm
    Clear
    Snap cap
    Pack of 100
    Pack of 100 - $55.83
    $55.83
    Pack of 100
       $55.83
     : 9502C-E-10C
  • PE
    Soft-Guard, White Silicone Rubber / Red PTFE
    11 mm
    Clear
    Snap cap
    Case of 1,000
    Case of 1,000 - $545.99
    $545.99
    Case of 1,000
       $545.99
     : 9502C-E-10C-M
  • PE
    Soft-Guard, White Silicone Rubber / Red PTFE
    11 mm
    Green
    Snap cap
    Pack of 100
    Pack of 100 - $55.83
    $55.83
    Pack of 100
       $55.83
     : 9502C-E-10G
  • PE
    Soft-Guard, White Silicone Rubber / Red PTFE
    11 mm
    Green
    Snap cap
    Case of 1,000
    Case of 1,000 - $530.20
    $530.20
    Case of 1,000
       $530.20
     : 9502C-E-10G-M
  • PE
    Soft-Guard, White Silicone Rubber / Red PTFE
    11 mm
    Black
    Snap cap
    Pack of 100
    Pack of 100 - $56.68
    $56.68
    Pack of 100
       $56.68
     : 9502C-E-10K
  • PE
    Soft-Guard, White Silicone Rubber / Red PTFE
    11 mm
    Black
    Snap cap
    Case of 1,000
    Case of 1,000 - $530.20
    $530.20
    Case of 1,000
       $530.20
     : 9502C-E-10K-M
  • PE
    Soft-Guard, White Silicone Rubber / Red PTFE
    11 mm
    Light Blue
    Snap cap
    Pack of 100
    Pack of 100 - $55.83
    $55.83
    Pack of 100
       $55.83
     : 9502C-E-10LB
  • PE
    Soft-Guard, White Silicone Rubber / Red PTFE
    11 mm
    Light Blue
    Snap cap
    Case of 1,000
    Case of 1,000 - $530.20
    $530.20
    Case of 1,000
       $530.20
     : 9502C-E-10LB-M
  • PE
    Soft-Guard, White Silicone Rubber / Red PTFE
    11 mm
    Red
    Snap cap
    Pack of 100
    Pack of 100 - $55.83
    $55.83
    Pack of 100
       $55.83
     : 9502C-E-10R
  • PE
    Soft-Guard, White Silicone Rubber / Red PTFE
    11 mm
    Red
    Snap cap
    Case of 1,000
    Case of 1,000 - $545.99
    $545.99
    Case of 1,000
       $545.99
     : 9502C-E-10R-M
  • PE
    Soft-Guard, White Silicone Rubber / Red PTFE
    11 mm
    Yellow
    Snap cap
    Pack of 100
    Pack of 100 - $56.68
    $56.68
    Pack of 100
       $56.68
     : 9502C-E-10Y
  • PE
    Soft-Guard, White Silicone Rubber / Red PTFE
    11 mm
    Yellow
    Snap cap
    Case of 1,000
    Case of 1,000 - $530.20
    $530.20
    Case of 1,000
       $530.20
     : 9502C-E-10Y-M
  • PE
    Soft-Guard, White Silicone Rubber / Red PTFE
    11 mm
    Blue
    Snap cap
    Pack of 100
    Pack of 100 - $57.49
    $57.49
    Pack of 100
       $57.49
     : 9502C-E-10B
Snap Top, 11 mm
  • Matière
    Septum
    Taille du bouchon
    Cap Color
    Cap Type
  • PE
    White Silicone Rubber / Natural PTFE
    11 mm
    Blue
    Snap cap
    Pack of 100
    Pack of 100 - $57.73
    $57.73
    Pack of 100
       $57.73
    Minimum order quantity is 1
     : 9502C-E-15B
  • PE
    White Silicone Rubber / Natural PTFE
    11 mm
    Blue
    Snap cap
    Case of 100
    Case of 100 - $349.12
    $349.12
    Case of 100
       $349.12
    Minimum order quantity is 1
     : 9502C-E-15B-M
  • PE
    White Silicone Rubber / Natural PTFE
    11 mm
    Clear
    Snap cap
    Pack of 100
    Pack of 100 - $48.38
    $48.38
    Pack of 100
       $48.38
    Minimum order quantity is 1
     : 9502C-E-15C
  • PE
    White Silicone Rubber / Natural PTFE
    11 mm
    Clear
    Snap cap
    Case of 1,000
    Case of 1,000 - $397.26
    $397.26
    Case of 1,000
       $397.26
    Minimum order quantity is 1
     : 9502C-E-15C-M
  • PE
    White Silicone Rubber / Natural PTFE
    11 mm
    Green
    Snap cap
    Pack of 100
    Pack of 100 - $48.38
    $48.38
    Pack of 100
       $48.38
    Minimum order quantity is 1
     : 9502C-E-15G
  • PE
    White Silicone Rubber / Natural PTFE
    11 mm
    Green
    Snap cap
    Case of 1,000
    Case of 1,000 - $349.12
    $349.12
    Case of 1,000
       $349.12
    Minimum order quantity is 1
     : 9502C-E-15G-M
  • PE
    White Silicone Rubber / Natural PTFE
    11 mm
    Black
    Snap cap
    Pack of 100
    Pack of 100 - $45.57
    $45.57
    Pack of 100
       $45.57
    Minimum order quantity is 1
     : 9502C-E-15K
  • PE
    White Silicone Rubber / Natural PTFE
    11 mm
    Black
    Snap cap
    Case of 1,000
    Case of 1,000 - $344.03
    $344.03
    Case of 1,000
       $344.03
    Minimum order quantity is 1
     : 9502C-E-15K-M