Screw Caps, MicroSolv™
Supplier: MicroSolv Technology Corp
Jump to:
À propos de cet article
These blue screw caps are designed for use with 9-425, 12x32 mm vials.
6 Options Available Below
Product Details & Documents
These blue screw caps are designed for use with 9-425, 12x32 mm vials.
Product Family Options
Informations sur le produit
- MatièreSeptumTaille du bouchonCap ColorCap TypeAvailabilityPrice
- PPBonded Silicone/PTFE, low durometer9-425 mmBlueScrew cap, knurledLog in pour la disponibilitéPack of 100$79.93Supplier Number: 9502S-10C-B
- PPBonded Silicone/PTFE, low durometer9-425 mmBlueScrew cap, knurledLog in pour la disponibilitéCase of 1,000$755.13Supplier Number: 9502S-10C-B-M
- PPBonded Silicone/PTFE, pre-slit9-425 mmBlueScrew cap, knurledLog in pour la disponibilitéPack of 100$99.16Supplier Number: 9502S-30C-B
- PPBonded Silicone/PTFE, pre-slit9-425 mmBlueScrew cap, knurledLog in pour la disponibilitéCase of 1,000$928.84Supplier Number: 9502S-30C-B-M
- PPBonded Silicone/PTFE9-425 mmBlueScrew cap, knurledLog in pour la disponibilitéPack of 100$135.81Supplier Number: 9502S-16B
- PPFitted Silicone/PTFE9-425 mmBlueScrew cap, smooth sided, Robo StyleLog in pour la disponibilitéCase of 1,000$635.18Supplier Number: 9502S-05-B-M