VWR® Storage Carts | Avantor
VWR® Storage Carts
VWR® Storage Carts
Supplier:  VWR International
Certificates
VWR® Storage Carts
VWR® Storage Carts
41 Options Available Below
Product Details & Documents
Storage carts made from food-grade, high-density polyethylene have four 3" industrial casters, two of which lock.
  • Locking hinged door
  • Bulk storage area with door
  • One key locks drawers and bulk storage
  • Full-extension drawers with handle
  • Polyethylene construction resists chemicals
The carts feature full-extension drawers for full viewing of drawer contents.
The carts add extra storage and work space in labs, clinics, hospitals, and other locations. They move all supplies together in a single work area, so no one needs to make extra trips to set up an activity.
They create storage for glassware, chemicals, pens, pipets, tools, tweezers, bottles, and other supplies. The top-side shelves are a built-in dispenser for Kimwipes and gloves. Shelves snap in and out as needed. All drawers and bulk storage areas lock with provided keys.
Ordering information: Five flexible storage carts include 1, 2, 3, or 4 full-extension drawers, 1 large bulk storage area, 4 or 6 side shelves, and 2 built-in dispensers for gloves and wipes. The carts are available in five sizes and multiple colors. A total-replacement lifetime manufacturer's guarantee is provided. A rail top for security is optional.
When ordering a custom printed cart the pantone color or vector of logo/image must be submitted. Be sure to include the location of logo/image.
Product Family Options
Informations sur le produit
  • Désignation
    Color
    No. of Drawers
    Weight
    Availability
    Price
  • PL22 mini storage cart
    Beige
    1
    37 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,700.44
    $3,700.44
    Prix promotionnel pour les nouveaux clients
    Each
       $3,700.44
    La quantité minimale de commande est de 1
  • PL27 small storage cart
    Beige
    2
    44 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,988.20
    $3,988.20
    Prix promotionnel pour les nouveaux clients
    Each
       $3,988.20
    La quantité minimale de commande est de 1
  • PL31 medium storage cart
    Beige
    2
    48 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,495.81
    $4,495.81
    Each
       $4,495.81
    La quantité minimale de commande est de 1
  • PL36 large storage cart
    Beige
    3
    56 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,638.88
    $4,638.88
    Each
       $4,638.88
    La quantité minimale de commande est de 1
  • PL36 large storage cart, rail top
    Beige
    3
    56 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5,111.15
    $5,111.15
    Each
       $5,111.15
    La quantité minimale de commande est de 1
  • PL22 mini storage cart
    Turqouise
    1
    37 lbs.
    Discontinued
  • PL27 small storage cart
    Turqouise
    2
    44 lbs.
    Discontinued
  • PL31 medium storage cart
    Turqouise
    2
    48 lbs.
    Discontinued
  • PL36 large storage cart
    Turqouise
    3
    56 lbs.
    Discontinued
  • PL41 jumbo storage cart
    Turqouise
    4
    63 lbs.
    Discontinued
  • PL27 small storage cart, rail top
    Turqouise
    2
    44 lbs.
    Discontinued
  • PL31 medium storage cart, rail top
    Turqouise
    2
    48 lbs.
    Discontinued
  • PL36 large storage cart, rail top
    Turqouise
    3
    56 lbs.
    Discontinued
  • PL22 mini storage cart
    Black
    1
    37 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,864.79
    $3,864.79
    Prix promotionnel pour les nouveaux clients
    Each
       $3,864.79
    La quantité minimale de commande est de 1
  • PL27 small storage cart
    Black
    2
    44 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,079.29
    $4,079.29
    Prix promotionnel pour les nouveaux clients
    Each
       $4,079.29
    La quantité minimale de commande est de 1
  • PL31 medium storage cart
    Black
    2
    48 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,347.77
    $4,347.77
    Each
       $4,347.77
    La quantité minimale de commande est de 1
  • PL36 large storage cart
    Black
    3
    56 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,687.54
    $4,687.54
    Each
       $4,687.54
    La quantité minimale de commande est de 1
  • PL41 jumbo storage cart
    Black
    4
    63 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,028.60
    $6,028.60
    Each
       $6,028.60
    La quantité minimale de commande est de 1
  • PL22 mini storage cart, rail top
    Black
    1
    37 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,003.66
    $4,003.66
    Prix promotionnel pour les nouveaux clients
    Each
       $4,003.66
    La quantité minimale de commande est de 1
  • PL36 large storage cart, rail top
    Black
    3
    56 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,796.74
    $4,796.74
    Each
       $4,796.74
    La quantité minimale de commande est de 1
  • PL36 large storage cart, custom
    3
    56 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $7,069.97
    $7,069.97
    Each
       $7,069.97
    La quantité minimale de commande est de 1
  • PL41 jumbo storage cart, custom
    4
    63 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $8,802.52
    $8,802.52
    Each
       $8,802.52
    La quantité minimale de commande est de 1
  • PL36 large storage cart, custom rail top
    3
    56 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5,060.22
    $5,060.22
    Prix promotionnel pour les nouveaux clients
    Each
       $5,060.22
    La quantité minimale de commande est de 1
  • PL27 small storage cart
    Blue
    2
    44 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,080.49
    $4,080.49
    Prix promotionnel pour les nouveaux clients
    Each
       $4,080.49
    La quantité minimale de commande est de 1
  • PL31 medium storage cart
    Blue
    2
    48 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,503.11
    $4,503.11
    Each
       $4,503.11
    La quantité minimale de commande est de 1
  • PL36 large storage cart
    Blue
    3
    56 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,652.69
    $4,652.69
    Each
       $4,652.69
    La quantité minimale de commande est de 1
  • PL41 jumbo storage cart
    Blue
    4
    63 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5,672.24
    $5,672.24
    Each
       $5,672.24
    La quantité minimale de commande est de 1
  • PL27 small storage cart, rail top
    Blue
    2
    44 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,180.73
    $4,180.73
    Prix promotionnel pour les nouveaux clients
    Each
       $4,180.73
    La quantité minimale de commande est de 1
  • PL22 mini storage cart, rail top
    Blue
    1
    37 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,796.64
    $3,796.64
    Prix promotionnel pour les nouveaux clients
    Each
       $3,796.64
    La quantité minimale de commande est de 1
  • PL31 medium storage cart, rail top
    Blue
    2
    48 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,436.56
    $4,436.56
    Each
       $4,436.56
    La quantité minimale de commande est de 1
  • PL36 large storage cart, rail top
    Blue
    3
    56 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5,111.15
    $5,111.15
    Each
       $5,111.15
    La quantité minimale de commande est de 1
  • PL27 small storage cart
    Lavender
    2
    44 lbs.
    Discontinued
  • PL31 medium storage cart, flat top
    Lavender
    2
    48 lbs.
    Discontinued
  • PL36 large storage cart
    Lavender
    3
    56 lbs.
    Discontinued
  • PL41 jumbo storage cart, rail top
    Lavender
    4
    63 lbs.
    Discontinued
  • PL22 mini storage cart
    Silver
    1
    37 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,864.79
    $3,864.79
    Prix promotionnel pour les nouveaux clients
    Each
       $3,864.79
    La quantité minimale de commande est de 1
  • PL27 small storage cart
    Silver
    2
    44 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,157.20
    $4,157.20
    Prix promotionnel pour les nouveaux clients
    Each
       $4,157.20
    La quantité minimale de commande est de 1
  • PL31 medium storage cart
    Silver
    2
    48 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,478.16
    $4,478.16
    Each
       $4,478.16
    La quantité minimale de commande est de 1
  • PL36 large storage cart
    Silver
    3
    56 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,780.24
    $4,780.24
    Each
       $4,780.24
    La quantité minimale de commande est de 1
  • PL27 small storage cart, rail top
    Silver
    2
    44 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,248.59
    $4,248.59
    Prix promotionnel pour les nouveaux clients
    Each
       $4,248.59
    La quantité minimale de commande est de 1
  • PL31 medium storage cart, rail top
    Silver
    2
    48 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,377.30
    $4,377.30
    Each
       $4,377.30
    La quantité minimale de commande est de 1
  • PL36 large storage cart, rail top
    Silver
    3
    56 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,874.58
    $4,874.58
    Each
       $4,874.58
    La quantité minimale de commande est de 1
  • PL41 jumbo storage cart, rail top
    Silver
    4
    63 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5,874.54
    $5,874.54
    Each
       $5,874.54
    La quantité minimale de commande est de 1
  • PL22 mini storage cart
    White
    1
    37 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,476.68
    $3,476.68
    Prix promotionnel pour les nouveaux clients
    Each
       $3,476.68
    La quantité minimale de commande est de 1
  • PL27 small storage cart
    White
    2
    44 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,934.42
    $3,934.42
    Prix promotionnel pour les nouveaux clients
    Each
       $3,934.42
    La quantité minimale de commande est de 1
  • PL31 medium storage cart
    White
    2
    48 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,253.79
    $4,253.79
    Prix promotionnel pour les nouveaux clients
    Each
       $4,253.79
    La quantité minimale de commande est de 1
  • PL36 large storage cart
    White
    3
    56 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5,199.15
    $5,199.15
    Each
       $5,199.15
    La quantité minimale de commande est de 1
  • PL41 jumbo storage cart
    White
    4
    63 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5,604.02
    $5,604.02
    Each
       $5,604.02
    La quantité minimale de commande est de 1
  • PL22 mini storage cart, rail top
    White
    1
    37 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,818.22
    $3,818.22
    Prix promotionnel pour les nouveaux clients
    Each
       $3,818.22
    La quantité minimale de commande est de 1
  • PL27 small storage cart, rail top
    White
    2
    44 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,197.16
    $4,197.16
    Prix promotionnel pour les nouveaux clients
    Each
       $4,197.16
    La quantité minimale de commande est de 1
  • PL31 medium storage cart, rail top
    White
    2
    48 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,101.62
    $2,101.62
    Prix promotionnel pour les nouveaux clients
    Each
       $2,101.62
    La quantité minimale de commande est de 1
  • PL36 large storage cart, rail top
    White
    3
    56 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5,255.70
    $5,255.70
    Each
       $5,255.70
    La quantité minimale de commande est de 1
  • PL41 jumbo storage cart, rail top
    White
    4
    63 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,154.48
    $6,154.48
    Each
       $6,154.48
    La quantité minimale de commande est de 1