Dispersion Blade Impellers | Avantor
Dispersion Blade Impellers
Supplier:   Caframo
Dispersion Blade Impellers
Supplier:  Caframo
Certificates
Dispersion Blade Impellers
Dispersion Blade Impellers
3 Options Available Below
Product Details & Documents
Caframo Dispersion Blade Impellers are electropolished 316 stainless steel dispersion blades ideal for high shear mixing at medium to high speeds, include blade only.
  • Designed with a uniform bore (8 mm) compatible with all Caframo shafts
  • Available in multiple sizes
  • Quick changes to test and optimize different batch sizes
  • Chemically inert and corrosion resistant
  • Broken particles are efficiently dispersed
When mixing, particles hit the blades and are broken apart and dispersed by the laminar flow set up by the blade.
52428-778 features a 1.9" diameter with 5/16" bore, and includes 4 set screws and Allen key. 
Ordering information: Shaft available separately.
Delivery information: Supplied with set screws.
Product Family Options
Informations sur le produit
  • Désignation
    Dimensions
    Matière
    Ø
    Availability
    Price
  • Dispersion blade impeller for radial flow
    Bore: 8 mm (⁵⁄₁₆")
    Electropolished 316 stainless steel
    48 mm (1.9")
    Discontinued
    Supplier Number: A164
  • Dispersion blade impeller for radial flow
    Bore: 8 mm (⁵⁄₁₆")
    Electropolished 316 stainless steel
    48 mm (1.9")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $496.82
    $496.82
    Each
       $496.82
    La quantité minimale de commande est de 1
    Supplier Number: A164
  • Dispersion blade impeller for radial flow
    Bore: 8 mm (⁵⁄₁₆")
    Electropolished 316 stainless steel
    75 cm (3")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $359.00
    $359.00
    Each
       $359.00
    Supplier Number: A431
  • Dispersion blade impeller for radial flow
    Electropolished 316 stainless steel
    100 mm (4")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $390.01
    $390.01
    Each
       $390.01
    Supplier Number: A441