Pivoting Blade Impeller, 2.4", Caframo
Fournisseur: Caframo
Certificats
À propos de cet article
The Caframo Pivoting Blade Impeller features a 2.4" diameter, and is pinned to a 5/16" by 16" shaft
The impeller will vortex 5 gallons of water at 800rpm. Designed for use with narrow neck vessels.
Composed of 316L electropolished stainless steel, it offers moderate shear with moderate vortexing characteristics.
Frequently Bought Together