Kevorkian Biopsy Punch, OR Grade, Sklar
10194-070
: Sklar
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Length:292 mm (11½")
- Material:Stainless steel
- Cat. no.:10194-070
- Shape:Straight
- Supplier No.:90-7540
Kevorkian Biopsy Punch is used during colposcopy procedures to excise tissue from the endocervix for biopsy.
This product has a 3 x 8-mm rectangular bite with 4 lower teeth. There is a basket to catch the tissue sample after excision. The non-ratcheted, spring style handle features one large ring that can fit multiple fingers and has a horn on the upper handle for thumb placement.