To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Sterility:Non sterile
- Description:Tischler Oval Biopsy Punch
- Material:Stainless steel
- Tip Configuration:Elliptical / Oval Bite with 1 Lower Tooth
- Curvature:Straight
- Cat. no.:10193-958
- Supplier No.:90-7520
Spécifications
À propos de cet article
Sklar's® Tischler Elliptical / Oval Biopsy Punch is a non-ratcheted, double spring OB/GYN instrument used in colposcopy procedures to obtain cervical tissue samples for biopsy.
- 0.3 x0.7 cm Bite
- Oval shaped tissue sample
- Pistol style handle with one large ring for multiple fingers
- Censitrac Ready products are industry standard and constructed of stainless steel
- Reusable
- Latex-free
It features a single, lower tooth used to grasp difficult to reach tissue and a locking mechanism above the horn on the handle so the jaws can lock the tissue sample into place while the instrument is removed from the cervical canal
A standard size biopsy punch may produce cause more bleeding and pain than a mini or baby punch; however, with certain diagnoses it is necessary to obtain a larger tissue sample. The length of the cannula/tip is approximately 8".
This reduces the risk of losing the tissue sample as the instrument is withdrawn.