Staticide® ESD Hi-Tech Hand Lotion, ACL
Catalogue # 75780-064
Fournisseur: ACL Staticide
Spécifications
- Capacity:473 ml (16 oz.)
- Description:Staticide® ESD Hi-Tech Hand Lotion with Pump
- Cat. no.:75780-064
- Supplier No.:7016
Spécifications
À propos de cet article
Static-dissipative hand lotion is essential for replenishing vital moisture on the skin that is important for wrist strap performance
This static-dissipative hand lotion is equipped to prevent and handle static discharge, especially in the electronics industry.
Certifications: Complies with DOD 1686A and DOD HDBK 263.