To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Sterility:Non sterile
- Description:Micro-Cut Tischler Oval Biopsy Punch with Locking Tip
- Material:Stainless steel
- Tip Configuration:Elliptical/Oval Bite; 1 Lower Tooth
- Curvature:Straight
- Cat. no.:10183-848
- Supplier No.:90-8940
Sklar's® Micro-Cut Tischler Biopsy Punch Forceps are an OB/GYN instrument most commonly used in colposcopy procedures to obtain cervical tissue samples for biopsy.
- 0.3 x 0.7 cm Bite
- Oval shaped tissue sample
- Pistol style handle with one large ring for multiple fingers
- Reusable
- Latex-free
It features a single, lower tooth used to grasp difficult to reach tissue and a locking mechanism above the horn on the handle so the jaws can lock the tissue sample into place while the instrument is removed from the cervical canal
Censitrac Ready OR Grade Micro-Cut Tischler Oval Biopsy Punch is industry standard and constructed of stainless steel.
This reduces the risk of losing the tissue sample as the instrument is withdrawn. A standard size biopsy punch may produce greater bleeding and pain than a mini or baby punch.