Precision Suture Scissors, OR Grade, Sklar
Catalogue # 89025-316
Fournisseur: Sklar
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Length:89 mm (3½")
- Material:Stainless steel
- Cat. no.:89025-316
- Supplier No.:22-2822
- Blade Type:Curved, Sharp/Sharp
Spécifications
À propos de cet article
Precision Suture Scissors are used for suture removal
This instrument has sharp tips, that easily slide under and gently lift sutures in preparation for removal. One of the sharp/sharp blades has a notch that the suture rests in before it is cut.