ContactUsLinkComponent
Surgi-OR™ TC Halsey Needle Holders, Mid Grade, Sklar®
Catalogue # 89023-490
Fournisseur:  Sklar
undefined
Surgi-OR™ TC Halsey Needle Holders, Mid Grade, Sklar®
Catalogue # 89023-490
Fournisseur:  Sklar
Supplier Number:  95-133
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
  • Length:
    133 mm (5¼")
  • Material:
    Tungsten carbide/stainless steel
  • Cat. no.:
    89023-490
  • Jaw Type:
    Cross-serrated
  • Tip Style:
    Straight
  • Supplier No.:
    95-133
Spécifications
À propos de cet article
Surgi-OR™ TC Halsey Needle Holders are used for holding small needles for suturing, most commonly during plastic surgery and procedures with delicate tissue, such as dermatology.
They are available with smooth or delicately serrated jaws. Smooth jaws will cause the least damage to the needle; however, serrated jaws hold the needle more tightly. Physician preference typically determines the type of working surface utilized. Surgi-OR™ TC mid-grade products are reliable instrumentation for the operating room and other surgical centers.
Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision.