HyClone Characterized Fetal Bovine Serum, Canadian origin, uses "True Pool" processing technology to ensure consistency in the quality of serum.
- Assayed for a variety of critical components and biochemical characteristics
- Endotoxin-tested: certificate of analysis confirms ≤25 EU/ml endotoxin
- Post-filtration treatments
The final serum is assayed for provides important proteins, hormones, growth factors, metabolites, and nutrients for that support your healthy cell culture process. Assay results are included in the certificate of analysis and biochemical assay list.
Gamma-irradiated and heat-inactivated FBS options are available to further reduce the risk of adventitious agents, particularly when manufacturing products for use in humans or animals. The irradiation process is validated to deliver a minimum dose of 25 kGy.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- CA95059-636
- CA76236-334
- CA76236-336
- CA95059-634
- Endotoxin:
- ≤25 EU/ml
- ≤25 EU/ml
- ≤25 EU/ml
- ≤25 EU/ml
- Hemoglobin:
- ≤25 mg/dl
- ≤25 mg/dl
- ≤25 mg/dl
- ≤25 mg/dl
- Storage temperature:
- ‒10 °C
- ‒10 °C
- ‒10 °C
- ‒10 °C