Acinetobacter Leeds Medium, Hardy Diagnostics | Avantor
Acinetobacter Leeds Medium, Hardy Diagnostics
Acinetobacter Leeds Medium, 18 mL
Catalog # 10068-018
Supplier:   Hardy Diagnostics
Fournisseur #:  G261
Acinetobacter Leeds Medium, Hardy Diagnostics
Catalog # 10068-018
Supplier:  Hardy Diagnostics
CAS Number:  
Acinetobacter Leeds Medium, Hardy Diagnostics
Acinetobacter Leeds Medium, Hardy Diagnostics
Acinetobacter Leeds Medium, 18 mL
CAS Number:  
Price
$89.40
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents
Leeds Acinetobacter Medium is a selective and differential medium intended for use as a plating technique to obtain a pure culture of Acinetobacter.
  • Selective components inhibit bacterial growth
  • Circular, convex, smooth and opaque colonies
  • Selective, differential medium
The selective components in Leeds Acinetobacter Medium are traditionally cefsulodin and cephradine, which inhibit the growth of non-Acinetobacter gram-negative bacteria, and vancomycin, which inhibits the growth of gram-positive bacteria
Acinetobacter spp. will produce light pink mucoid colonies with a pink to mauve color diffused into the medium. The colonies are circular, convex, smooth, and opaque with entire margins of 1 to 2 mm in diameter after 24 hours at 35°C.
The presence of fructose and sucrose and neutral red, a pH indicator, allows the differentiation of bacteria based on the products released when the bacteria use fructose and sucrose as a nutrient source. The colonies of those that release acidic products are yellow, while the colonies of those that release alkaline products are red.
Spécifications
Spécifications
  • Désignation:
    Acinetobacter Leeds Medium
  • Taille:
    18 mL