À propos de cet article
Sklar Blue™ Auvard Weighted Vaginal Speculum is a single end vaginal speculum commonly used in electrosurgical OB/GYN procedures.
Sklar Blue™ electrosurgical instruments feature high quality stainless steel with an insulated coating. This safe coating prevents radio frequency currents from being transmitted through the instrument which reduces the burning of non-targeted tissue. Sklar Blue instruments are fully autoclavable and may also be cleaned ultrasonically.
It is available with various blades styles and three weight choices in order to accommodate the needs of the patient and the procedure. The weight holds the speculum in place and provides an open view for the surgeon to perform procedures.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.