À propos de cet article
Pierce™ Trypsin Protease, MS Grade, is highly purified porcine trypsin that has been chemically modified for maximum activity and stability in proteomic applications.
- Cleaves at the carboxyl side of lysine and arginine residues with greater than 95% specificity
- No detectable chymotrypsin activity thus making it highly pure
- Chemically modified for reduced autolytic activity
Trypsin is a serine protease derived from porcine pancreatic extracts. Applications include in-gel digestion of proteins from 1-D or 2-D gels and in-solution tryptic digestion of proteins. The selectivity of this enzyme is critical for reproducible protein digestion and mass spectrometry-based protein identification. It is TPCK-treated to eliminate chymotryptic activity and methylated to improve stability during protein digestion. The enzyme is most active in pH ranges 7 to 9 and can be reversibly inactivated at pH <4.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Spécifications
- Storage conditions:Upon receipt, store at -20˚C in a nonfrost-free freezer. Products are shipped with ice.