ContactUsLinkComponent
Pierce™ Endoproteinase Lys-C Protease, MS Grade, Thermo Scientific
Catalog # CAPI90051
Supplier:  Thermo Scientific
CAS Number:  
undefined
Pierce™ Endoproteinase Lys-C Protease, MS Grade, Thermo Scientific
Catalog # CAPI90051
Supplier:  Thermo Scientific
Supplier Number:  90051
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Endoproteinase Lys-C protease, MS grade
  • Size:
    1 x 20 µg
  • Cat. no.:
    CAPI90051
  • Supplier No.:
    90051
Specifications
About this item
Endoproteinase Lys-C protease is a highly purified native endoproteinase validated for maximum activity and stability in proteomic applications.
  • Can be used in tandem with trypsin to decrease tryptic missed cleavages thus enhancing digestion
  • Better protein characterization results from overlapping peptides with complementary chromatographic, ionisation and fragmentation properties thereby increasing sequence coverage
  • Effective enzyme activity under highly denaturing conditions
  • C-terminal lysine cleavage specificity of 90% for a complex protein sample
  • Stable since it is provided in a lyophilized format
This Lys-C is a mass spectrometry (MS)-grade serine protease isolated from Lysobacter enzymogenes
Applications include improved sequence coverage of protein digests, de novo sequencing, epigenetic studies and in-gel and in-solution digestion of proteins.
Lys-C has high activity and specificity for lysine residues resulting in larger peptides and less sample complexity than trypsin (i.e. fewer peptides). Unlike trypsin, Lys-C can cleave lysines followed by prolines, making it ideal for sequential protein digestion followed by trypsin to decrease missed cleavages. These unique Lys-C properties ensure high digestion efficiency when used alone or followed by tryptic digestion.
Caution: For Research Use Only. Not for use in diagnostic procedures.