À propos de cet article
B-PER™ Complete Bacterial Protein Extraction Reagent is an all-in-one formulation of non ionic detergent and enzymes that enables mild extraction of proteins from both gram-negative and gram-positive bacteria without the need for mechanical disruption. B-PER Complete Reagent combines a universal nuclease and lysozyme to improve cell lysis and reduce viscosity, and may be used for soluble protein extraction and inclusion body purification from bacterial cell lysates.
- Fast and simple - just add B-PER Reagent to a bacterial pellet, incubate with mixing for 10 to 15 minutes and recover soluble proteins after pelleting the cell debris
- Convenient - contains lysozyme and a universal nuclease in a single formulation with 4°C storage
- Excellent yields - recover recombinant proteins from bacterial lysates or purify inclusion bodies to near-homogeneous levels
- Suitable for any scale of protein extraction
- Completely compatible with addition of protease inhibitors, and the resulting protein extract can be used in protein assays, typical affinity purification methods (eg. GST, 6xHis) and other applications
B-PER™ Complete Bacterial Protein Extraction Reagents are compatible with downstream applications, such as affinity chromatography (eg. immobilised metal affinity chromatography, glutathione chromatography), SDS-PAGE, and protein quantification. Depending on the particular application, protease inhibitors, salts, reducing agents, denaturants, and chelating agents may be added to the reagent.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Spécifications
- Storage conditions:Store at 4˚C