ContactUsLinkComponent
Pierce™ Primary Cardiomyocyte Isolation Kit, Thermo Scientific
Pierce™ Primary Cardiomyocyte Isolation Kit, Thermo Scientific
Catalog # CAPI88281
Supplier:  Thermo Scientific
CAS Number:  
undefined
Pierce™ Primary Cardiomyocyte Isolation Kit, Thermo Scientific
Catalog # CAPI88281
Supplier:  Thermo Scientific
Supplier Number:  88281
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Primary Cardiomyocyte Isolation Kit
  • Size:
    1 L
  • Cat. no.:
    CAPI88281
  • Supplier No.:
    88281
  • Sufficient For:
    50 Neonatal Mouse/Rat Hearts
Specifications
About this item
The Pierce™ Primary Cardiomyocyte Isolation Kit provides isolation and culturing reagents for the optimal yield and viability of primary cardiomyocytes from neonatal heart from mouse and rat.
  • Procedure and reagents optimized for viability, yield, purity, and ease-of-use
  • Isolation protocol requires less than 2 hours compared to up to 21 hours by other methods
  • Provides a 1.5- to 4-fold increase in yield compared to do-it-yourself methods and other commercial kits
  • Provides higher viability than do-it-yourself methods and other commercial kits
  • Cultured cardiomyocytes express the appropriate biochemical markers with validated beating activity
The Pierce™ Primary Cardiomyocyte Isolation Kit has been designed to obtain high viability and purity of primary cardiomyocytes using validated reagents and an optimized procedure for enzymatic digestion and culturing. The kit contains two tissue-specific dissociation enzymes, media formulated for primary cell culture, and a cardiomyocyte cell culture growth supplement that promotes the isolation, enrichment and growth of functional cardiomyocytes. The validated protocol ensures optimal isolation and significantly reduces the processing time of isolation when compared to other methods. Addition of the cardiomyocyte growth supplement to culture cells further enriches for cardiomycytes, and cultures have been maintained for up to 7 days using the kit components.
Ordering information: Upon receipt store Product 88281X at 4?C and Product 88281Y at -20?C. Product 88281X is shipped on ice packs and Product 88281Y is shipped with dry ice.
Delivery: Kit includes Cardiomyocyte Isolation Enzyme 1, Cardiomyocyte Isolation Enzyme 2, Hanks' Balanced Salt Solution (HBSS), D-MEM for Primary Cell Isolation, and Cardiomyocyte Growth Supplement.
Caution: For Research Use Only. Not for use in diagnostic procedures.