Order Entry
Canada
ContactUsLinkComponent
 
Pierce™ Mouse Embryonic Fibroblast Isolation Kit, Thermo Scientific
Catalog # CAPI88279
Supplier:  Thermo Scientific
CAS Number:  
undefined
Pierce™ Mouse Embryonic Fibroblast Isolation Kit, Thermo Scientific
Catalog # CAPI88279
Supplier:  Thermo Scientific
Supplier Number:  88279
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Mouse embryonic fibroblast isolation kit
  • Size:
    1 L
  • Cat. no.:
    CAPI88279
  • Supplier No.:
    88279
  • Sufficient For:
    Fibroblast Isolation from 50 Embryonic Mouse Somatic Tissues

Specifications

About this item

The Pierce™ Mouse Embryonic Fibroblast Isolation Kit provides isolation and culturing reagents for the optimal yield and viability of fibroblasts from mouse embryos.

  • Procedure and reagents optimized for viability, yield, purity, and ease-of-use
  • Provides a 2-fold increase in yield compared to do-it-yourself methods and other kits
  • Cultured fibroblasts express the appropriate biochemical markers

"MEF's isolated and cultured using the Pierce™ MEF Isolation Kit can be used as a cell culture model for a diverse range of studies, such as gene regulation and stem cell research. MEF's are used most commonly as feeder layers to maintain mouse embryonic stem cells in an undifferentiated state. With a combination of transcription factors, MEFs can be converted to a pluripotent state or directly reprogrammed to various types of cells, such as functional neurons and cardiomyocytes, which could have important implications for studies of development, neurological and cardiac disease modelling, drug discovery, and regenerative medicine. The Pierce™ MEF Isolation Kit also can be used to isolate MEF's from different genetically altered mouse models to study growth control and DNA damage response. MEF's isolated and cultured with the Pierce™ MEF Isolation Kit can be readily frozen for future use with minimal loss of viability.

Delivery: Kit includes MEF Isolation Enzyme, Hanks' balanced salt solution (HBSS), and D-MEM for Primary Cell Isolation.
Caution: For Research Use Only. Not for use in diagnostic procedures.