ContactUsLinkComponent
Pierce™ IgM Purification Kit, Thermo Scientific
undefined
Pierce™ IgM Purification Kit, Thermo Scientific
  • Storage conditions:
    Sufficient For: 10 IgM purifications, each from 0.5 mL mouse ascites samples
    MBP Agarose Column, 5 mL, 1 column
    IgM Binding Buffer, 800 mL
    IgM Elution Buffer, 500 mL
    MBP Column Preparation Buffer, 50 mL
    Column Extender
    Store at 4˚C
  • Shipping temperature:
    ambient
Frequently Bought Together