Pierce™ StartingBlock™ (PBS) Blocking Buffer, Thermo Scientific
Pierce™ StartingBlock™ (PBS) Blocking Buffer, Thermo Scientific
Pierce™ StartingBlock™ (PBS) Blocking Buffer, Thermo Scientific
Pierce™ StartingBlock™ (PBS) Blocking Buffer, Thermo Scientific
These StartingBlock™ blocking buffers are PBS and TBS formulations of a proprietary protein that provides exceptional performance and broad compatibility with membrane (Western blot) and 96-well plate (ELISA) assays.
  • Compatible with many detection systems (Western blot, ELISA and IHC)
  • Short blocking times
  • High signal:noise ratio
  • Strip and reprobe without reblocking
  • Available ready to use 1X formulations in PBS and TBS
StartingBlock Blocking Buffers performs well with an extremely wide range of antibodies, antibody combinations, other protein probing and assay systems.
These buffers are biotin- and serum-free. They are compatible with many detection systems and feature short blocking times. These buffers also allow stripping and reprobing with reblocking to save time and money.
 : For Research Use Only. Not for use in diagnostic procedures. Upon receipt store at 4˚C.
Product Family Options
  • Désignation
    Taille
    Utilisation
  • PBS
    1 L
    ELISA, Western Blot, Dot Blot, Immunohistochemistry
    Each
    Each - $623.43
    $623.43
    Each
       $623.43
     : 37538
  • PBS with Tween® 20
    1 L
    ELISA, Western Blot, Dot Blot, Immunohistochemistry
    Each
    Each - $664.76
    $664.76
    Each
       $664.76
     : 37539
  • TBS
    1 L
    ELISA, Western Blot, Dot Blot, Immunohistochemistry
    Each
    Each - $634.33
    $634.33
    Each
       $634.33
     : 37542
  • TBS with Tween® 20
    1 L
    ELISA, Western Blot, Dot Blot, Immunohistochemistry
    Each
    Each - $675.75
    $675.75
    Each
       $675.75
     : 37543
  • Blocking Buffer
    100 mL
    ELISA, Western Blot, Dot Blot, Immunohistochemistry
    Each (100ml)
    Each (100ml) - $140.26
    $140.26
    Each (100ml)
       $140.26
     : 37578
  • Blocking Buffer
    100 mL
    ELISA, Western Blot, Dot Blot, Immunohistochemistry
    Each (100ml)
    Each (100ml) - $139.87
    $139.87
    Each (100ml)
       $139.87
     : 37579