ContactUsLinkComponent
Pierce™ 1-Step™ Ultra TMB-Blotting Solution, Thermo Scientific
Catalogue # CAPI37574
Fournisseur:  Thermo Scientific
Numéro CAS:  
undefined
Pierce™ 1-Step™ Ultra TMB-Blotting Solution, Thermo Scientific
Catalogue # CAPI37574
Fournisseur:  Thermo Scientific
Supplier Number:  37574
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    1-Step™ Ultra TMB-Blotting Solution
  • Size:
    250 mL
  • Cat. no.:
    CAPI37574
  • Supplier No.:
    37574
Spécifications
À propos de cet article
Thermo Scientific 1-Step Ultra TMB-Blotting Solution is an enhanced single-component horseradish peroxidase (HRP) substrate for Western blotting and immunohistochemistry.
  • Protein bands visible in less than one minute
  • Protein bands stable after membrane drying
  • No special equipment needed for visualization, produces dark blue bands
  • Ready to use and sensitive
This precipitating, colorimetric Western blot substrate for HRP provides high sensitivity, increased signal-to-noise ratio and low background compared to many other chromogenic substrates. The blotting solution contains soluble TMB (3,3',5,5'-tetramethylbenzidine), which reacts very quickly with horseradish peroxidase enzyme to produce an insoluble dark blue precipitate that does not fade or flake. The substrate is compatible with both nitrocellulose and PVDF membranes.
Caution: For Research Use Only. Not for use in diagnostic procedures. Upon receipt store product at 4˚C.