To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Description:Compat-Able™ Protein Assay Reagent Kit
- Size:500 mL
- Cat. no.:CAPI23215
- Supplier No.:23215
The Compat-Able™ Protein Assay Preparation Reagent Set eliminates interfering substances from protein assay samples to make them compatible with BCA and Bradford assays.
- Removes nearly any substance that interferes with colorimetric protein assays: salts, detergents, denaturants, reducing agents, and even IEF sample buffer
- Prepare samples in less than 10 minutes with four simple steps
- Reagent set is easier to use than homemade TCA or acetone precipitation reagents, thereby producing more consistent results
- Microcentrifuge method is easily scaled to larger centrifuge tube volumes for special needs
- Protocol integrates control and normalization procedures
The Compat-Able Reagents remove salts, detergents, reducing agents and other substances from protein samples to eliminate interference with protein assays. The protocol selectively precipitates all of the protein, thereby allowing the non-protein components to be easily decanted and removed. The purified, precipitated protein is redissolved in water, making it ready for accurate determination of the protein concentration by standard protein assay methods. The procedure is accomplished in less than 10 minutes with just a few simple steps.