Order Entry
Canada
ContactUsLinkComponent
Pierce™ Monomeric Avidin Agarose Kit, Thermo Scientific
Pierce™ Monomeric Avidin Agarose Kit, Thermo Scientific
Catalog # CAPI20227
Supplier:  Thermo Scientific
CAS Number:  
undefined
Pierce™ Monomeric Avidin Agarose Kit, Thermo Scientific
Catalog # CAPI20227
Supplier:  Thermo Scientific
Supplier Number:  20227
CAS Number:  

Specifications

  • Description:
    Pierce™ Monomeric Avidin Agarose Kit
  • Size:
    2 mL
  • Cat. no.:
    CAPI20227
  • Supplier No.:
    20227
  • Sufficient For:
    Binding ≥2,4 mg biotinylated BSA per column

Specifications

About this item

The Thermo Scientific Pierce™ Monomeric Avidin Kit enables mild affinity purification of biotinylated proteins, peptides and other molecules

  • Monomeric Avidin Agarose can be regenerated and reused at least 10 times
  • Retains biotin-binding specificity and low nonspecific binding of native avidin
  • Bind samples in a variety of physiological buffers
  • Binding capacity >1.2 mg biotinylated BSA/mL resin

During monomeric avidin immobilization, polymeric forms of avidin with strong binding characteristics also are immobilized. These high affinity biotin-binding sites must first be blocked with a biotin-containing buffer. Biotin molecules are eluted from monomers with a glycine solution revealing only the reversible binding sites. The biotinylated molecule of interest may then be applied to the support for purification and then eluted by ligand competition using a biotin-containing solution.

Avidin monomers have a much lower biotin-binding affinity than native tetrameric avidin enabling dissociation of biotinylated molecules using mild elution conditions

Delivery: Kit includes Pierce™ Monomeric Avidin column, 2 mL (1 column), PBS pack, makes 500 mL (1 pack), Biotin Blocking and Elution Buffer (200 mL), Regeneration Buffer (250 mlL, and column accessories.
Caution: For Research Use Only. Not for use in diagnostic procedures.