ContactUsLinkComponent
Pierce™ LightShift™ Chemiluminescent RNA EMSA Kit, Thermo Scientific
  CAPI20158
 :  
undefined
Pierce™ LightShift™ Chemiluminescent RNA EMSA Kit, Thermo Scientific
  CAPI20158
 :  20158
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    LightShift™ Chemiluminescent RNA EMSA Kit
  • Cat. no.:
    CAPI20158
  • For Membrane Area:
    1000 cm² (10 mini gel blots)
  • No. of Reactions:
    100
  • Supplier No.:
    20158
The Thermo Scientific LightShift™ Chemiluminescent RNA EMSA Kit provides a non-radioactive solution for studying RNA-protein interactions using an electrophoretic mobility-shift assay (EMSA).
  • Chemiluminescent detection comparable to radioactive detection
  • Develop X-ray films after 1 to 5 minute exposures thus being a time saving process
  • Compatible with RNA probes biotinylated by different methods
  • Complete kit includes optimized reagents for binding reactions and RNA probe detection
  • Eliminate waste concerns from radioactive RNA probes
Chemiluminescent RNA EMSA Kit is an in vitro technique for detection of protein-RNA interactions through changes in gel electrophoresis migration patterns. This kit uses biotinylated RNA probes and a chemiluminescent substrate system to achieve fast and safe detection of RNA-protein complexes with sensitivity equivalent to traditional 32P-isotopic methods.
 : Kit includes 125 nM Biotinylated IRE RNA control (35 µL), 10 µM unlabeled IRE RNA control (25 µL), 2 mg/mL Cytosolic liver extract (50 µL), 10 mg/mL tRNA (100 µL), 10X REMSA binding buffer (1 mL), 50% Glycerol (500 µL), 2M KCl (500 µL), 1M MgCl2 (500 µL), DTT lyophilized, nuclease-free water (1,5 mL), 5X REMSA loading buffer (1 mL), Stabilized streptavidin-HRP (1,5 mL), Luminol/enhancer solution (80 mL), Stable peroxide solution (80 mL), Nucleic acid detection blocking buffer (500 mL), 4X wash buffer (500 mL), and Substrate equilibration buffer (500 mL).
 : For Research Use Only. Not for use in diagnostic procedures.