To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:LightShift™ Chemiluminescent RNA EMSA Kit
- Cat. no.:CAPI20158
- For Membrane Area:1000 cm² (10 mini gel blots)
- No. of Reactions:100
- Supplier No.:20158
Specifications
About this item
The Thermo Scientific LightShift™ Chemiluminescent RNA EMSA Kit provides a non-radioactive solution for studying RNA-protein interactions using an electrophoretic mobility-shift assay (EMSA).
- Chemiluminescent detection comparable to radioactive detection
- Develop X-ray films after 1 to 5 minute exposures thus being a time saving process
- Compatible with RNA probes biotinylated by different methods
- Complete kit includes optimized reagents for binding reactions and RNA probe detection
- Eliminate waste concerns from radioactive RNA probes
Chemiluminescent RNA EMSA Kit is an in vitro technique for detection of protein-RNA interactions through changes in gel electrophoresis migration patterns. This kit uses biotinylated RNA probes and a chemiluminescent substrate system to achieve fast and safe detection of RNA-protein complexes with sensitivity equivalent to traditional 32P-isotopic methods.