Formaldehyde Solution Spill Kit | Avantor
Formaldehyde Solution Spill Kit
Formaldehyde Solution Spill Kit
Catalog # 470219-190
Supplier:   Ward's Science
Fournisseur #:  IS5035
Formaldehyde Solution Spill Kit
Catalog # 470219-190
Supplier:  Ward's Science
Formaldehyde Solution Spill Kit
Formaldehyde Solution Spill Kit
Formaldehyde Solution Spill Kit
Price
$104.60
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
Product Details & Documents
Be prepared for spills in your lab.
  • Enough Material to Clean One Liter of a 10% Formalin Solution
  • Instructions and SDS
  • Vinyl Exam Gloves
This kit includes a dust pan and brush to properly dispose of spills.
It also includes vinyl exam gloves, so you are protected while cleaning.
Ordering information: This kit includes an instruction sheet/SDS, vinyl exam gloves, polybags, twist ties, blank shipping tags, dust pan and brush, vermiculite, and an absorbent formaldehyde neutralizing agent (sodium bisulfite).
Spécifications
  • Désignation:
    Formaldehyde Solution Spill Kit