Two-Piece Tissue Cassettes
Product Details & Documents
Two-Piece Tissue Cassettes are molded from a special, high-density polymer that is resistant to the chemical actions of solvents.
- Three frosted writing areas
- Two-piece design
- Lid relocks without risk of specimen loss
- Chemical-resistant
Lids can be snapped on and off as often as necessary and relock without danger of specimen loss.
Cassettes have a two-piece design and feature three frosted labeling areas.
: 500 per case.
Documents
Spécifications
- Color:White
- Désignation:Two-piece tissue cassettes