PLUGGO™ Decappers, LPG Consulting | Avantor
PLUGGO™ Decappers, LPG Consulting
PLUGGO Standard Decapper Carousel, 24-Place, 110 - 120 V, 60 Hz, 32 W, Vacuum Collection Tubes, 10 x 8 x 16", 13 x 75 mm, 13 x 100 mm, 16 x 75 mm and 16 x 100 mm
Catalog # CA12103-000
Supplier:   LGP Consulting
Fournisseur #:  4600 4450
PLUGGO™ Decappers, LPG Consulting
Catalog # CA12103-000
Supplier:  LGP Consulting
PLUGGO™ Decappers, LPG Consulting
PLUGGO™ Decappers, LPG Consulting
PLUGGO Standard Decapper Carousel, 24-Place, 110 - 120 V, 60 Hz, 32 W, Vacuum Collection Tubes, 10 x 8 x 16", 13 x 75 mm, 13 x 100 mm, 16 x 75 mm and 16 x 100 mm
Price
48 703,05 $
Log in pour la disponibilité
Each
Product Details & Documents
PLUGGO decapper assemblies and rack handlers are designed for the safe removal of original stoppers from plastic or glass vacuum tubes.
The PLUGGO SOLO single tube decapper is an automated benchtop decapper for a single tube. It occupies a small benchtop footprint and can be used with glass or plastic tubes. The PLUGGO Standard decapper is a carousel that can process up to 50 tubes per minute.
The PLUGGO RHs (Rack Handler for single rack) handles a single rack at a time, with room to stage additional racks. The PLUGGO RH (Rack Handler for multiple racks) handles up to 15 racks in and out. The RHs and the RH are available for Abbott Architect, Beckman AU, Iris/Sysmex and Roche analyzer racks.
The systems significantly reduce the risk of infection through aerosol formation or accidental blood spillage, particularly with the use of vacuum blood collection tubes. The risk of repetitive strain injury is also greatly reduced due to the simplicity of the system's operation.
Spécifications
  • Désignation:
    PLUGGO Standard Decapper Carousel, 24-Place
  • W×D×H:
    10 x 8 x 16"
  • Electrical:
    110 - 120 V, 60 Hz, 32 W
  • For Use With:
    Vacuum Collection Tubes
  • Tube Size:
    13 x 75 mm, 13 x 100 mm, 16 x 75 mm and 16 x 100 mm
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Replacement Carousels for 13 x 75 mm Tubes
    Replacement Carousels for 13 x 75 mm Tubes
    CA12103-002
    Log in pour la disponibilité
    Each - $329.22
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Replacement Carousels for 13 x 100 mm Tubes
    Replacement Carousels for 13 x 100 mm Tubes
    CA12103-004
    Log in pour la disponibilité
    Each - $334.39
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Replacement Carousels for 16 x 100 mm Tubes
    Replacement Carousels for 16 x 100 mm Tubes
    CA12103-006
    Log in pour la disponibilité
    Each - $334.39
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Replacement Carousels for 16 x 75 mm Tubes
    Replacement Carousels for 16 x 75 mm Tubes
    CA12103-008
    Log in pour la disponibilité
    Each - $328.55