Phosphate Buffers, Epredia™
Supplier:   Thermo Scientific
Phosphate Buffers, Epredia™
Supplier:  Thermo Scientific
Certificates
Phosphate Buffers, Epredia™
1 Options Available Below
Product Details & Documents
Epredia™ Phosphate Buffers, ready-to-use dry powder for a variety of hematology staining techniques.
  • Buffer supplied as 9.25 g powder
Product Family Options
Informations sur le produit
  • Désignation
    pH
    Taille
    Availability
    Price
  • Phosphate Buffer
    6.6
    9.25 g (0.34 oz.)
    Discontinued
    Supplier Number: 89032
  • Phosphate Buffer
    7.2
    9.25 g (0.34 oz.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 12
    Case of 12 - $1,639.18
    $1,639.18
    Case of 12
       $1,639.18
    Supplier Number: 89033