Lab Worktables, Stationary, Metro™
Supplier: Metropolitan Wire
Certificates
About this item
Stainless steel lab worktables provides corrosion-proof performance. Stainless steel provides excellent protection in corrosive environments.
- Tables feature smooth, easy-to-clean surfaces
- Tops are made of Type 304 stainless steel or Trespa® phenolic resin
- Phenolic resin tops are compression molded of organic fiber-reinforced phenolic core with an integrally cured thermoset resin surface
- Phenolic resin tops are ideal for moderate to high chemical use and have excellent abrasion resistance and impact resistance
- Tops with backsplash feature a 10.2 cm (4") high backsplash
Stainless lab tables are load rated at 50 lbs. per square foot, up to a maximum of 600 lbs. when load is evenly distributed and caster specifications meet requirements. Tables are 30" deep.
Frequently Bought Together