ContactUsLinkComponent
ELIMINASE®, Decontaminants, Decon Labs
Catalog # 10798-804
Supplier:  Decon Labs
CAS Number:  
undefined
ELIMINASE®, Decontaminants, Decon Labs
Catalog # 10798-804
Supplier:  Decon Labs
Supplier Number:  1101
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Pack type:
    12/case
  • Description:
    ELIMINase
  • Size:
    250 ml
  • Cat. no.:
    10798-804
  • Supplier No.:
    1101
Specifications
About this item
ELIMINase removes RNase, DNase, unwanted DNA and radioactive contamination.
  • Phosphate free and non-toxic
  • Safely disposable and totally rinsable for the decontaminated surface
  • Eliminates the time-consuming method of autoclaving
It works instantly so there is no waiting. ELIMINase is an ideal substitute for using DiEthylPryoCarbonate (DEPC) which is a carcinogen. ELIMINase can be used on all surfaces including glass, plastic, metal and ceramic.