ContactUsLinkComponent
Forensic Analysis of Fabrics and Fibers
Catalogue # 470219-556
Fournisseur:  Ward's Science
undefined
Forensic Analysis of Fabrics and Fibers
Catalogue # 470219-556
Fournisseur:  Ward's Science
Supplier Number:  IS9018
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Forensic Analysis of Fabrics and Fibers
  • No. of Lab Groups:
    10
  • Cat. no.:
    470219-556
  • Includes:
    Teacher ‘s Guide and Student Copymasters
  • Supplier No.:
    IS9018
Spécifications
À propos de cet article
What Can You Learn from Fabric?
  • Learn forensic techniques used to analyze fabrics
  • Discover how a fabric's physical and chemical characteristics can be unique
This kit has been designed to introduce students to the concepts of fabric and fiber analysis. Experiments in this kit cover the areas of cloth weave identification, microscopic fiber analysis, burn testing and chemical analysis. Students will first analyze a group of known materials to learn the various techniques. They will then be given a crime scene sample and need to test materials taken from four suspects to try and identify who might be involved.
Materials Included:
Acetone
Sodium Hypochlorite (Bleach)
Hydrochloric Acid
Sodium Hydroxide
Mounting Medium
pH Strips
Microscope Slides and Coverslips
Lead Acetate Paper
Metal Forceps
Cotton Fabric Sample
Wool Fabric Sample
Acetate Fabric Sample
Polypropylene Fabric Sample
Polyester Fabric Sample
Crime Scene Sample
Suspect Samples #1-4