Cryogenic Printer Labels, Brady
À propos de cet article
Cryogenic labels for tubes, vials, and canes. Chemical- and stain-resistant labels for slides and cassettes. General labels for straws, petri dishes, bottles, flasks, microplates and bags.
Cryogenic labels: temperatures from –196 to 121 °C (–321 to 250 °F) and hot water baths.
B-461 Ultra-thin label with clear tail for tubes.
B-490 Freezerbondz® label for frozen tubes.
B-492 Freezerbondz® ultra-thin for frosted/frozen tubes and canes.
General labels:
B-424 Economical paper label for general purpose.
B-427 Straw label for wrapping; working temperatures from –196 to 80 °C (–321 to 176 °F).
B-430 Petri dish and bag label; working temperatures from –80 to 121 °C (–112 to 250 °F).
B-432 Removable label for all room temperature applications.
4 Options Available Below
Product Details & Documents
Cryogenic labels for tubes, vials, and canes. Chemical- and stain-resistant labels for slides and cassettes. General labels for straws, petri dishes, bottles, flasks, microplates and bags.
Cryogenic labels: temperatures from –196 to 121 °C (–321 to 250 °F) and hot water baths.
B-461 Ultra-thin label with clear tail for tubes.
B-490 Freezerbondz® label for frozen tubes.
B-492 Freezerbondz® ultra-thin for frosted/frozen tubes and canes.
General labels:
B-424 Economical paper label for general purpose.
B-427 Straw label for wrapping; working temperatures from –196 to 80 °C (–321 to 176 °F).
B-430 Petri dish and bag label; working temperatures from –80 to 121 °C (–112 to 250 °F).
B-432 Removable label for all room temperature applications.
Product Family Options
B-492 Freezerbondz® Ultra-Thin for Frosted/Frozen Tubes and Canes
Informations sur le produit
- DésignationMatièreColorPour l'utilisation avecDimensionsPackagingAvailabilityPrice
- Cryogenic printer labelsPolyesterWhiteBBP®339.5×25.4 mm (0.38×1")Log in pour la disponibilitéRoll of 1,500Roll of 1,500 - $275.32$275.32Prix promotionnel pour les nouveaux clientsEach$275.32Roll of 1,500$275.32Supplier Number: B33-163-492
- Cryogenic printer labelsPolyesterWhiteBBP®3312.7×25.4 mm (0.5×1")Discontinued-Supplier Number: -
- Cryogenic printer labelsPolyesterWhiteBBP®3312.7×25.4 mm (0.5×1")Log in pour la disponibilitéRoll of 1,500Roll of 1,500 - $351.01$351.01Prix promotionnel pour les nouveaux clientsEach$351.01Roll of 1,500$351.01Supplier Number: B33-182-492
- Cryogenic printer labelsPolyesterWhiteBBP®339.5×25.4 mm (0.38×1")Log in pour la disponibilitéRoll of 2,500Roll of 2,500 - $441.67$441.67Prix promotionnel pour les nouveaux clientsEach$441.67Roll of 2,500$441.67Supplier Number: B33-152-492
- Cryogenic printer labelsPolyesterWhiteBBP®3325.4×48.3 mm (1×1.9")Discontinued-Supplier Number: -
B-423 Product and Component Identification Labels, PCB and Rating Plates
Informations sur le produit
- DésignationMatièreColorPour l'utilisation avecDimensionsPackagingAvailabilityPrice
- Printer labelsPolyesterWhiteBBP®336,4×44,5 mm (0.25×1.75")1 label/rowLog in pour la disponibilitéRoll of 1$165.08Supplier Number: 133961