Sodium Cacodylate Buffer, Electron Microscopy Sciences
Sodium Cacodylate Buffer, Electron Microscopy Sciences
Sodium Cacodylate Buffer, Electron Microscopy Sciences
Certificates
Sodium Cacodylate Buffer, Electron Microscopy Sciences
Sodium Cacodylate Buffer, Electron Microscopy Sciences
7 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Used for preparing buffered glutaraldehdye fixatives for electron microscopy
Available with a pH of 7.2 or 7.4.
Sodium Cacodylate Buffer, 0.2M is prepared from Sodium Cacodylate Trihydrate (F.W 214.02).
Product Family Options
Product Information
  • Désignation
    pH
    Taille
    Availability
    Price
  • Sodium Cacodylate Buffer, 0.2M
    Specify
    225 ml
    Log in for availability
    Log in for availability
    Each (225ml)
    Each (225ml) - $702.54
    $702.54
    Each (225ml)
       $702.54
    Supplier Number: 11650
  • Sodium Cacodylate Buffer, 0.2M
    Specify
    500 ml
    Log in for availability
    Log in for availability
    Each (500ml)
    Each (500ml) - $1,502.44
    $1,502.44
    Each (500ml)
       $1,502.44
    Supplier Number: 11652
  • Sodium Cacodylate Buffer, 0.2M
    Specify
    500 ml
    Log in for availability
    Log in for availability
    Each (500ml)
    Each (500ml) - $146.54
    $146.54
    Each (500ml)
       $146.54
    Supplier Number: 11652
  • Sodium Cacodylate Buffer, 0.2M
    Specify
    225 ml
    Log in for availability
    Log in for availability
    Each (225ml)
    Each (225ml) - $105.10
    $105.10
    Each (225ml)
       $105.10
    Supplier Number: 11650
  • Sodium Cacodylate Buffer, 0.2M
    7.4
    1 L
    Log in for availability
    Log in for availability
    Each
    Each - $287.64
    $287.64
    Each
       $287.64
    Supplier Number: 11653
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Sodium Cacodylate Buffer, 0.4M
    7.2
    500 ml
    Log in for availability
    Log in for availability
    Each
    Each - $213.69
    $213.69
    Each
       $213.69
    Supplier Number: 11654
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Sodium Cacodylate Buffer, 0.4M
    7.2
    1 L
    Log in for availability
    Log in for availability
    Each
    Each - $377.11
    $377.11
    Each
       $377.11
    Supplier Number: 11655
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.