Combined Hematoxylin and Eosin/Methenamine Silver Stain for the Histological Diagnosis of Fungi in Tissue, Electron Microscopy Sciences
Catalogue # CA11028-246
Fournisseur: Electron Microscopy Science
Numéro CAS:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Description:Chromic Acid 10%
- Cat. no.:CA11028-246
- Supplier No.:26123-01
Spécifications
À propos de cet article
A Combined Hematoxylin and Eosin/Methenamine Silver Stain is used for the Histological Diagnosis of Fungi in Tissue Sections.
- For bacterial, fungal stains
Stain results - Organism: Blue-Black Background: Rose.
Lamps, Laura W. et al A Combined Hematoxylin and Eosin/Methenamin Silver Stain for theHistological Diagnosis of Fungi In Tissue Sections., J. of Histotechnology 23:4 p.341 – 345, 2000.