Tyrode Solution, Electron Microscopy Sciences
Tyrode Solution, Electron Microscopy Sciences
Certificates
1 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Specifications for Product Family
Spécifications
Spécifications
  • Catalog No:
  • 100496-406
  • 100502-380
  • Size:
  • 500 ml
Spécifications
Product Family Options
Informations sur le produit
  • Désignation
    Type de conditionnement
    Taille
    Availability
    Price
  • Tyrode Solution
    Bottle
    500 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (500ml)
    Each (500ml) - $78.02
    $78.02
    Each (500ml)
       $78.02
    Supplier Number: 11760-05
    Spécifications:
    Minority or Woman-Owned Business Enterprise
    More
    Specifications
    Cat. No.
    100496-406
    Size
    500 ml
    Minority or Woman-Owned Business Enterprise
  • Tyrode Solution
    Bottle
    1 l
    Discontinued
    Supplier Number: 11760-10
    Spécifications:
    Minority or Woman-Owned Business Enterprise
    More
    Specifications
    Cat. No.
    100502-380
    Size
    Minority or Woman-Owned Business Enterprise